Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mob4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133528.1 | NP_598212.1 | 7 | 495 | ||
GenBank mRNA | AJ132008.1 | - | 7 | 495 | ||
BC085708.1 | - | 7 | 516 | 7193103 | ||
FQ229796.1 | - | 7 | 511 |