Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd79b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133533.2 | NP_598217.1 | 5, 6 | 655 | ||
GenBank mRNA | BC088115.1 | - | 5, 6 | 655 | 7325406 | |
FQ226266.1 | - | 5, 6 | 652 | |||
FQ227061.1 | - | 5, 6 | 650 | |||
FQ227788.1 | - | 5, 6 | 647 | |||
FQ230261.1 | - | 5, 6 | 646 | |||
FQ232054.1 | - | 5, 6 | 646 | |||
FQ232680.1 | - | 5, 6 | 646 | |||
FQ232790.1 | - | 5, 6 | 669 | |||
FQ233507.1 | - | 5, 6 | 644 | |||
FQ234082.1 | - | 5, 6 | 629 | |||
FQ234487.1 | - | 5, 6 | 644 |