Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sult1c2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133547.4 | NP_598231.3 | 9 | 952 | ||
XM_006244242.2 | XP_006244304.1 | 8 | 928 | |||
XM_006244243.2 | XP_006244305.1 | 8 | 920 | |||
XM_006244245.2 | XP_006244307.1 | 8 | 915 | |||
XM_006244246.2 | XP_006244308.1 | 8 | 903 | |||
XM_006244248.2 | XP_006244310.1 | 9 | 1045 | |||
XM_006244249.2 | XP_006244311.1 | 7 | 854 | |||
XM_006244250.2 | XP_006244312.1 | 7 | 963 | |||
XM_008766738.1 | XP_008764960.1 | 8 | 933 | |||
XM_008766739.1 | XP_008764961.1 | 8 | 896 | |||
XM_008766740.1 | XP_008764962.1 | 8 | 926 | |||
XM_008766741.1 | XP_008764963.1 | 8 | 921 | |||
XM_008766742.1 | XP_008764964.1 | 8 | 908 | |||
XM_008766743.1 | XP_008764965.1 | 8 | 951 | |||
XM_008766744.1 | XP_008764966.1 | 7 | 841 | |||
XM_008766745.1 | XP_008764967.1 | 7 | 829 | |||
GenBank mRNA | AJ238391.1 | - | 6 | 734 | ||
BC103636.1 | - | 9 | 952 | 7106590 |
Gene Symbol: LOC100910057 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006244225.2 | XP_006244287.1 | 7 | 880 | ||
XM_006244227.2 | XP_006244289.1 | 8 | 917 | |||
XM_006244228.2 | XP_006244290.1 | 8 | 1222 | |||
XM_006244229.2 | XP_006244291.1 | 8 | 912 | |||
XM_006244230.2 | XP_006244292.1 | 8 | 1217 | |||
XM_006244231.2 | XP_006244293.1 | 7 | 858 | |||
XM_006244232.2 | XP_006244294.1 | 7 | 838 | |||
XM_006244233.2 | XP_006244295.1 | 7 | 1143 | |||
XM_006244236.2 | XP_006244298.1 | 5 | 576 | |||
XM_008766781.1 | XP_008765003.1 | 8 | 917 | |||
XM_008766782.1 | XP_008765004.1 | 8 | 900 | |||
XM_008766783.1 | XP_008765005.1 | 8 | 895 | |||
XM_008766784.1 | XP_008765006.1 | 8 | 873 | |||
XM_008766785.1 | XP_008765007.1 | 8 | 917 | |||
XM_008766786.1 | XP_008765008.1 | 8 | 917 |
Gene Symbol: LOC100910526 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006244254.2 | XP_006244316.1 | 763 |
Gene Symbol: RGD1562392 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003750614.3 | XP_003750662.1 | 7 | 861 | ||
XM_008766791.1 | XP_008765013.1 | 7 | 840 |