Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sult1c2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133547.4 | NP_598231.3 | 5 | 425 | ||
XM_006244242.2 | XP_006244304.1 | 4 | 401 | |||
XM_006244243.2 | XP_006244305.1 | 4 | 393 | |||
XM_006244245.2 | XP_006244307.1 | 4 | 388 | |||
XM_006244246.2 | XP_006244308.1 | 4 | 376 | |||
XM_006244248.2 | XP_006244310.1 | 5 | 518 | |||
XM_006244249.2 | XP_006244311.1 | 3 | 327 | |||
XM_006244250.2 | XP_006244312.1 | 3 | 436 | |||
XM_008766738.1 | XP_008764960.1 | 4 | 406 | |||
XM_008766739.1 | XP_008764961.1 | 4 | 369 | |||
XM_008766740.1 | XP_008764962.1 | 4 | 399 | |||
XM_008766741.1 | XP_008764963.1 | 4 | 394 | |||
XM_008766742.1 | XP_008764964.1 | 4 | 381 | |||
XM_008766743.1 | XP_008764965.1 | 4 | 424 | |||
XM_008766744.1 | XP_008764966.1 | 3 | 314 | |||
XM_008766745.1 | XP_008764967.1 | 3 | 302 | |||
GenBank mRNA | AJ238391.1 | - | 2 | 207 | ||
BC103636.1 | - | 5 | 425 | 7106590 |
Gene Symbol: LOC100910526 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006244254.2 | XP_006244316.1 | 236 |