Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dscam | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133587.1 | NP_598271.1 | 19 | 4016 | ||
GenBank mRNA | AF334385.1 | - | 19 | 4016 |
Set Membership: |
![]() |