Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lgals2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133599.1 | NP_598283.1 | 4 | 377 | ||
GenBank mRNA | AB001075.1 | - | 4 | 377 |