Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc31a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133600.1 | NP_598284.1 | 5 | 629 | ||
GenBank mRNA | AF268030.1 | - | 5 | 629 | ||
AY539951.1 | - | 4 | 461 | |||
BC078745.1 | - | 5 | 591 | 7126088 |