Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Khsrp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133602.1 | NP_598286.1 | 2 | 289 | ||
XM_006244292.2 | XP_006244354.1 | 2 | 427 | |||
GenBank mRNA | AF308818.1 | - | 2 | 289 |