Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc6a13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133623.1 | NP_598307.1 | 12 | 1449 | ||
XM_006237218.2 | XP_006237280.1 | 12 | 1466 | |||
XM_006237219.2 | XP_006237281.1 | 13 | 1582 | |||
XM_006237220.2 | XP_006237282.1 | 12 | 1448 | |||
XM_006237221.2 | XP_006237283.1 | 13 | 1564 | |||
XM_006237222.2 | XP_006237284.1 | 11 | 1320 | |||
XM_006237223.2 | XP_006237285.1 | 11 | 1447 | |||
GenBank mRNA | M95762.1 | - | 12 | 1449 |