Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: TERF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005652.4 | NP_005643.2 | 3 | 598 | ||
XM_005256121.3 | XP_005256178.1 | 3 | 614 | |||
XM_005256122.3 | XP_005256179.1 | 3 | 614 | |||
XM_005256123.3 | XP_005256180.1 | 3 | 614 | |||
XM_005256124.3 | XP_005256181.1 | 3 | 614 | |||
GenBank mRNA | AF002999.1 | - | 3 | 579 | ||
BC024890.1 | - | 3 | 581 | 4346819 | ||
U95970.1 | - | 3 | 337 |