Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TERF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005652.4 | NP_005643.2 | 7 | 1210 | ||
XM_005256121.3 | XP_005256178.1 | 8 | 1235 | |||
XM_005256122.3 | XP_005256179.1 | 8 | 1235 | |||
XM_005256123.3 | XP_005256180.1 | 7 | 1226 | |||
XM_005256124.3 | XP_005256181.1 | 5 | 992 | |||
GenBank mRNA | AF002999.1 | - | 7 | 1191 | ||
U95970.1 | - | 7 | 949 |