Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pou4f2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134355.1 | NP_599182.1 | 2 | 1653 | ||
XM_008772467.1 | XP_008770689.1 | 3 | 1251 |