Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FDFT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001287742.1 | NP_001274671.1 | 10 | 2330 | ||
NM_001287743.1 | NP_001274672.1 | 9 | 2132 | |||
NM_001287744.1 | NP_001274673.1 | 8 | 1959 | |||
NM_001287745.1 | NP_001274674.1 | 9 | 2038 | |||
NM_001287747.1 | NP_001274676.1 | 8 | 1995 | |||
NM_001287748.1 | NP_001274677.1 | 8 | 1657 | |||
NM_001287749.1 | NP_001274678.1 | 8 | 1895 | |||
NM_001287750.1 | NP_001274679.1 | 7 | 1897 | |||
NM_001287751.1 | NP_001274680.1 | 7 | 1769 | |||
NM_001287756.1 | NP_001274685.1 | 5 | 1354 | |||
NM_004462.4 | NP_004453.3 | 8 | 1841 | |||
XM_017013217.1 | XP_016868706.1 | 10 | 2622 | |||
GenBank mRNA | AK098682.1 | - | 1657 | |||
AK301617.1 | - | 5 | 1215 | |||
AK316534.1 | - | 7 | 1769 | |||
X69141.1 | - | 8 | 1710 |