Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prl8a9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134385.2 | NP_599212.1 | 2, 3 | 243 | ||
XM_006253904.1 | XP_006253966.1 | 2, 3 | 234 | |||
GenBank mRNA | AF239745.1 | - | 2, 3 | 243 | ||
AF239746.1 | - | 2, 3 | 240 | |||
AF239747.1 | - | 2, 3 | 243 | |||
BC100138.1 | - | 2, 3 | 236 | 7442617 | ||
BC127474.1 | - | 2, 3 | 234 | 7443385 |