Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Camk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134468.1 | NP_604463.1 | 10 | 1001 | ||
XM_006236999.2 | XP_006237061.1 | 11 | 1670 | |||
XM_006237000.2 | XP_006237062.1 | 11 | 1666 | |||
XM_006237001.2 | XP_006237063.1 | 10 | 1533 | |||
XM_006237002.2 | XP_006237064.1 | 10 | 1460 | |||
XM_006237003.2 | XP_006237065.1 | 10 | 1459 | |||
GenBank mRNA | BC071177.1 | - | 10 | 936 | 7098592 | |
FQ213485.1 | - | 964 | ||||
L24907.1 | - | 10 | 975 | |||
L26288.1 | - | 10 | 1001 |