Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp2c13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138514.1 | NP_612523.1 | 1 | 163 | ||
GenBank mRNA | BC088105.1 | - | 1 | 185 | 7372890 | |
BC098903.1 | - | 1 | 164 | 7373446 | ||
J02861.1 | - | 1 | 165 | |||
M32277.1 | - | 1 | 163 | |||
M33994.1 | - | 1 | 152 |