Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prok1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138851.1 | NP_620206.1 | 2, 3 | 205 | ||
GenBank mRNA | AY089983.1 | - | 2, 3 | 205 |