Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kat5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005872.1 | NP_001005872.1 | 6 | 613 | ||
GenBank mRNA | AF333984.1 | - | 4 | 266 | ||
BC083879.1 | - | 6 | 613 | 7130366 |