Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Opn4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138860.1 | NP_620215.1 | 1 | 324 | ||
GenBank mRNA | AY072689.1 | - | 1 | 324 |
Set Membership: |
![]() |