Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Spata7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138862.3 | NP_620217.2 | 7 | 945 | ||
XM_006240413.2 | XP_006240475.1 | 6 | 928 | |||
XM_006240414.2 | XP_006240476.1 | 7 | 1020 | |||
XM_006240415.2 | XP_006240477.1 | 5 | 853 | |||
XM_008764737.1 | XP_008762959.1 | 6 | 899 | |||
GenBank mRNA | BC078728.1 | - | 7 | 671 | 7114267 | |
CK470849.1 | - | 3 | 609 |