Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Svs3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007605.1 | NP_001007606.1 | 694 | |||
GenBank mRNA | D00920.1 | - | 642 |
Gene Symbol: LOC103691966 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008762551.1 | XP_008760773.1 | 2 | 866 |
Gene Symbol: LOC103694890 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008773871.1 | XP_008772093.1 | 1000 |