Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Svs3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007605.1 | NP_001007606.1 | 210 | |||
GenBank mRNA | D00920.1 | - | 158 |
Gene Symbol: LOC103690143 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008762549.1 | XP_008760771.1 | 2 | 516 |
Gene Symbol: LOC103691966 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008762551.1 | XP_008760773.1 | 2 | 382 |
Gene Symbol: LOC103694890 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008773871.1 | XP_008772093.1 | 516 |
Gene Symbol: Svs3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001102417.1 | NP_001095887.1 | 213 |