Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gls2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270786.1 | NP_001257715.1 | 7, 8 | 1078 | ||
NM_001270787.1 | NP_001257716.1 | 7, 8 | 1324 | |||
NM_138904.2 | NP_620259.2 | 6, 7 | 773 | |||
XM_006240728.2 | XP_006240790.1 | 7, 8 | 1104 | |||
XM_006240730.2 | XP_006240792.1 | 8, 9 | 903 | |||
GenBank mRNA | BC089776.1 | - | 7, 8 | 917 | ||
BC104712.1 | - | 7, 8 | 822 | |||
FQ211735.1 | - | 7, 8 | 1324 | |||
J05499.1 | - | 6, 7 | 772 |