Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Xab2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139109.2 | NP_620809.1 | 4 | 385 | ||
GenBank mRNA | AF277899.1 | - | 4 | 388 | ||
BC081723.1 | - | 4 | 379 | 7108549 |