Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Calcoco1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139190.1 | NP_631929.1 | 176 | |||
GenBank mRNA | AY078385.1 | - | 176 | |||
BC081722.1 | - | 146 | 7124320 |
Gene Symbol: LOC100912282 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003750418.3 | XP_003750466.1 | 2 | 240 |