Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plbd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139255.3 | NP_640348.2 | 9, 10 | 1284 | ||
GenBank mRNA | AY095481.1 | - | 3, 4 | 316 | ||
BC086408.1 | - | 4, 5 | 417 | |||
BC097934.1 | - | 5, 6 | 590 | |||
BC104715.1 | - | 4, 5 | 355 | |||
BC126078.1 | - | 5, 6 | 488 | |||
BC161835.1 | - | 5, 6 | 446 |