Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TGIF1 SIRNA Assays ›
Gene Symbol:
TGIF1
Gene Name:
TGFB induced factor homeobox 1
Gene Aliases:
HPE4, TGIF
Chromosome Location:
Chr.18: 3411927 - 3458412 on Build GRCh38
UniGene ID:
Hs.373550
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s14082
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TGIF1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001278682.1
NP_001265611.1
2
394
NM_001278684.1
NP_001265613.1
3
751
NM_001278686.1
NP_001265615.1
3
417
NM_003244.3
NP_003235.1
2
484
NM_170695.3
NP_733796.2
2
1128
NM_173207.2
NP_775299.1
2
362
NM_173208.2
NP_775300.1
3
754
NM_173209.2
NP_775301.1
2
570
NM_173210.2
NP_775302.1
2
521
NM_173211.1
NP_775303.1
2
233
NM_174886.2
NP_777480.1
3
551
XM_011525735.2
XP_011524037.1
2
730
XM_017025958.1
XP_016881447.1
3
815
XM_017025959.1
XP_016881448.1
2
600
GenBank mRNA
AF179900.1
-
2
172
AI813434.1
-
3
372
AK026766.1
-
1
1090
AK291112.1
-
2
318
AK309471.1
-
2
479
AU310737.1
-
2
484
BC000814.1
-
2
250
3453337
BC031268.1
-
2
856
5284849
BE296707.1
-
2
246
BE906361.1
-
3
403
BF244060.1
-
2
332
BF446044.1
-
3
366
BM849453.1
-
3
420
BQ437864.1
-
2
482
BU657748.1
-
2
338
BX952985.1
-
2
251
CB161219.1
-
2
394
CB996365.1
-
3
578
CR450333.1
-
2
166
D82120.1
-
2
233
DR007147.1
-
2
574
HY041619.1
-
2
363
X89750.1
-
2
477
Back To Top
More Information
Panther Classification:
Molecular Function -
homeodomain transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
transcription, DNA-templated
multicellular organism development
response to drug
cellular response to growth factor stimulus
RNA polymerase II core promoter proximal region sequence-specific DNA binding
transcriptional repressor activity, RNA polymerase II core promoter proximal region sequence-specific binding
transcription factor activity, sequence-specific DNA binding
transcription corepressor activity
protein binding
co-SMAD binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7c5f844ff9-f4tdd:80/10.239.165.3:80.
git-commit: 1d07a45fbd3445ee17db916b8ffd5a90ab36fdd0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.11.0-2024.07.28-1.0