Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prpf19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139333.3 | NP_647549.2 | 2 | 345 | ||
XM_006231013.2 | XP_006231075.1 | 2 | 381 | |||
XM_006231014.2 | XP_006231076.1 | 2 | 379 | |||
XM_006231015.2 | XP_006231077.1 | 2 | 380 | |||
GenBank mRNA | AB020022.2 | - | 2 | 391 | ||
BC107669.1 | - | 2 | 345 | 7937645 |