Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000360.3 | NP_000351.2 | 4 | 572 | ||
NM_199292.2 | NP_954986.2 | 5 | 665 | |||
NM_199293.2 | NP_954987.2 | 5 | 653 | |||
XM_011520335.2 | XP_011518637.1 | 4 | 2860 | |||
GenBank mRNA | BC104967.1 | - | 4 | 644 | 8143970 | |
BC143611.1 | - | 4 | 642 | 9052124 | ||
BC143614.1 | - | 4 | 656 | 9052127 | ||
BM718799.1 | - | 1 | 27 | |||
DQ677336.1 | - | 4 | 562 | |||
DQ677337.1 | - | 4 | 574 | |||
M17589.1 | - | 5 | 655 | |||
X05290.1 | - | 4 | 572 | |||
Y00414.1 | - | 5 | 647 |