Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: THBS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003247.3 | NP_003238.2 | 22 | 3744 | ||
GenBank mRNA | AK292429.1 | - | 22 | 3701 | ||
BC146676.1 | - | 21 | 3551 | 40148868 | ||
BC150175.1 | - | 21 | 3555 | 40147958 | ||
BX641023.1 | - | 8 | 1199 | |||
L12350.1 | - | 22 | 3663 | |||
M81339.1 | - | 11 | 1748 |