Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ctsz | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183330.1 | NP_899159.1 | 4 | 605 | ||
GenBank mRNA | AB023781.2 | - | 4 | 605 | ||
BC091110.1 | - | 4 | 539 | 7379777 |