Search Thermo Fisher Scientific
Gene Symbol: Hdgfrp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145785.1 | NP_665728.1 | 2 | 249 | ||
XM_008759496.1 | XP_008757718.1 | 2 | 524 | |||
GenBank mRNA | AF389347.1 | - | 2 | 249 |