Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other DOCK9 SIRNA Assays ›
Gene Symbol:
Dock9
Gene Name:
dedicator of cytokinesis 9
Gene Aliases:
Trg
Chromosome Location:
Chr.15: 107945664 - 108210985 on Build Rnor_6.0
UniGene ID:
Rn.10431
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID
s141310
Purification
Standard
HPLC
HPLC-IVR
Size
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
Availability
Made To Order
Catalog #
4390771
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Dock9
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001105759.2
NP_001099229.2
33, 34
3811
XM_006252481.2
XP_006252543.1
33, 34
3925
XM_006252488.2
XP_006252550.1
33, 34
3925
XM_006252490.2
XP_006252552.1
33, 34
3922
XM_006252493.2
XP_006252555.1
33, 34
4377
XM_006252496.2
XP_006252558.1
33, 34
3946
XM_006252499.2
XP_006252561.1
33, 34
3924
XM_006252501.2
XP_006252563.1
33, 34
3921
XM_006252502.2
XP_006252564.1
33, 34
3921
XM_006252503.2
XP_006252565.1
33, 34
3970
XM_006252505.2
XP_006252567.1
33, 34
3946
XM_008770951.1
XP_008769173.1
33, 34
3925
XM_008770952.1
XP_008769174.1
33, 34
3922
XM_008770953.1
XP_008769175.1
33, 34
3925
XM_008770954.1
XP_008769176.1
33, 34
3925
XM_008770955.1
XP_008769177.1
33, 34
3950
XM_008770956.1
XP_008769178.1
33, 34
3947
XM_008770957.1
XP_008769179.1
33, 34
3925
XM_008770958.1
XP_008769180.1
33, 34
3925
XM_008770959.1
XP_008769181.1
33, 34
3925
XM_008770960.1
XP_008769182.1
33, 34
3925
XM_008770961.1
XP_008769183.1
33, 34
3925
XM_008770962.1
XP_008769184.1
33, 34
3925
XM_008770963.1
XP_008769185.1
33, 34
3925
XM_008770964.1
XP_008769186.1
33, 34
3990
XM_008770965.1
XP_008769187.1
33, 34
3922
XM_008770966.1
XP_008769188.1
33, 34
3922
XM_008770967.1
XP_008769189.1
33, 34
3921
XM_008770968.1
XP_008769190.1
33, 34
4382
XM_008770969.1
XP_008769191.1
33, 34
4383
Back To Top
More Information
Panther Classification:
Molecular Function -
guanyl-nucleotide exchange factor
Gene Ontology Categories:
Function(s)
Process(es)
small GTPase mediated signal transduction
positive regulation of GTPase activity
guanyl-nucleotide exchange factor activity
Rho guanyl-nucleotide exchange factor activity
Rho GTPase binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-574f4745d9-7lstp:80/10.239.162.105:80.
git-commit: d20c60ba4e48417b3d41b4fa5195ecf3ea66aacf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.12.0-2024.07.30-1.0