Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Carhsp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152790.2 | NP_690003.1 | 3 | 227 | ||
XM_006245750.2 | XP_006245812.1 | 3 | 1010 | |||
XM_006245753.2 | XP_006245815.1 | 3 | 381 | |||
XM_008767479.1 | XP_008765701.1 | 3 | 704 | |||
GenBank mRNA | AF115346.1 | - | 2 | 197 | ||
BC071173.1 | - | 3 | 227 | 7099252 |