Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fzd9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153305.1 | NP_695217.1 | 1 | 1972 | ||
GenBank mRNA | AF455054.1 | - | 1 | 1972 | ||
DQ211688.1 | - | 1 | 1734 | |||
DQ211689.1 | - | 1 | 1734 |
Set Membership: |
![]() |