Gene Symbol: St18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153310.2 | NP_695222.2 | 18 | 2761 | ||
XM_008763489.1 | XP_008761711.1 | 18 | 3316 | |||
XM_008763490.1 | XP_008761712.1 | 19 | 3977 | |||
XM_008763491.1 | XP_008761713.1 | 18 | 3747 | |||
XM_008763492.1 | XP_008761714.1 | 18 | 3821 | |||
XM_008763493.1 | XP_008761715.1 | 17 | 2679 | |||
XM_008763494.1 | XP_008761716.1 | 14 | 2463 | |||
GenBank mRNA | AF031942.1 | - | 15 | 2585 | ||
U67080.1 | - | 18 | 2771 |