Gene Symbol: St18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153310.2 | NP_695222.2 | 12 | 2143 | ||
XM_008763489.1 | XP_008761711.1 | 12 | 2698 | |||
XM_008763490.1 | XP_008761712.1 | 13 | 3359 | |||
XM_008763491.1 | XP_008761713.1 | 12 | 3129 | |||
XM_008763492.1 | XP_008761714.1 | 12 | 3203 | |||
XM_008763493.1 | XP_008761715.1 | 11 | 2061 | |||
XM_008763494.1 | XP_008761716.1 | 8 | 1845 | |||
GenBank mRNA | AF031942.1 | - | 9 | 1967 | ||
U67080.1 | - | 12 | 2156 |