Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vom2r44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099498.1 | NP_001092968.1 | 4 | 1448 | ||
XM_006232438.2 | XP_006232500.1 | 4 | 2125 | |||
XM_008761031.1 | XP_008759253.1 | 4 | 1230 | |||
GenBank mRNA | AF053989.1 | - | 1 | 66 |
Set Membership: |
![]() |