Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Erc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_170788.2 | NP_740769.2 | 9 | 2098 | ||
XM_006237224.1 | XP_006237286.1 | 9 | 2098 | |||
XM_006237225.1 | XP_006237287.1 | 9 | 2098 | |||
XM_006237226.1 | XP_006237288.1 | 8 | 2014 | |||
XM_006237227.1 | XP_006237289.1 | 8 | 2014 | |||
XM_006237228.1 | XP_006237290.1 | 9 | 2098 | |||
XM_006237229.1 | XP_006237291.1 | 8 | 2014 | |||
XM_006237230.1 | XP_006237292.1 | 8 | 2014 | |||
XM_006237231.2 | XP_006237293.1 | 9 | 2098 | |||
XM_008763221.1 | XP_008761443.1 | 9 | 2098 | |||
XM_008763222.1 | XP_008761444.1 | 8 | 2014 | |||
XM_008763223.1 | XP_008761445.1 | 9 | 2098 | |||
GenBank mRNA | AF541926.1 | - | 9 | 2204 | ||
AY174115.1 | - | 9 | 2098 |