Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TKT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001064.3 | NP_001055.1 | 14 | 1942 | ||
NM_001135055.2 | NP_001128527.1 | 14 | 1942 | |||
NM_001258028.1 | NP_001244957.1 | 15 | 1966 | |||
NR_047580.1 | 14 | 1832 | ||||
XM_011534054.1 | XP_011532356.1 | 15 | 1904 | |||
XM_011534055.1 | XP_011532357.1 | 15 | 1991 | |||
GenBank mRNA | AK057092.1 | - | 8 | 1771 | ||
AK092084.1 | - | 9 | 2805 | |||
AK093667.1 | - | 13 | 1601 | |||
AK223614.1 | - | 14 | 1850 | |||
AK289454.1 | - | 14 | 1818 | |||
AK293438.1 | - | 15 | 1874 | |||
AK301231.1 | - | 14 | 1740 | |||
AK303191.1 | - | 15 | 1809 | |||
BC002433.2 | - | 15 | 1731 | 3346980 | ||
BC008615.1 | - | 9 | 1086 | |||
BC009970.2 | - | 14 | 1840 | 4310396 | ||
BC024026.2 | - | 12 | 1511 | |||
BQ430230.1 | - | 5 | 490 | |||
BX647248.1 | - | 14 | 1828 | |||
BX649193.1 | - | 6 | 7079 | |||
EU794667.1 | - | 14 | 1840 | |||
GQ472197.1 | - | 9 | 1086 | |||
KU178361.1 | - | 14 | 1770 | |||
KU178362.1 | - | 8 | 863 | |||
KU178363.1 | - | 7 | 656 | |||
L12711.1 | - | 14 | 1868 | |||
U55017.1 | - | 14 | 1770 | |||
X67688.1 | - | 15 | 1840 |