Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Rnf34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004075.1 | NP_001004075.1 | 2 | 222 | ||
XM_006249360.2 | XP_006249422.1 | 2 | 755 | |||
XM_006249361.2 | XP_006249423.1 | 3 | 432 | |||
XM_008769214.1 | XP_008767436.1 | 2 | 140 | |||
GenBank mRNA | AY157968.1 | - | 1 | 127 | ||
BC079044.1 | - | 2 | 222 | 7115027 |