Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TLE4 SIRNA Assays ›
Gene Symbol:
TLE4
Gene Name:
transducin like enhancer of split 4
Gene Aliases:
BCE-1, BCE1, E(spI), ESG, ESG4, GRG4, Grg-4
Chromosome Location:
Chr.9: 79571773 - 79726882 on Build GRCh38
UniGene ID:
Hs.444213
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s14176
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TLE4
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001282748.1
NP_001269677.1
4
1253
NM_001282749.1
NP_001269678.1
4
1253
NM_001282753.1
NP_001269682.1
4
1253
NM_001282760.1
NP_001269689.1
4
1253
NM_007005.4
NP_008936.2
4
1253
NR_104239.1
4
1253
XM_005252177.1
XP_005252234.1
4
426
XM_006717264.1
XP_006717327.1
4
426
XM_006717268.1
XP_006717331.1
4
426
XM_011518952.1
XP_011517254.1
4
426
XM_011518953.1
XP_011517255.1
4
303
XM_011518955.1
XP_011517257.1
4
426
XM_011518956.1
XP_011517258.1
4
426
XM_011518957.1
XP_011517259.1
4
426
XM_011518958.1
XP_011517260.1
4
426
XM_011518959.1
XP_011517261.1
4
426
XM_011518960.1
XP_011517262.1
4
426
XM_011518961.1
XP_011517263.1
4
426
XM_011518962.1
XP_011517264.1
4
426
XM_011518963.1
XP_011517265.1
4
426
XM_011518964.1
XP_011517266.1
4
426
XM_011518965.1
XP_011517267.1
4
426
XM_011518966.1
XP_011517268.1
4
426
XM_011518967.1
XP_011517269.1
4
426
XM_011518968.1
XP_011517270.1
4
426
XM_011518969.1
XP_011517271.1
4
426
XM_011518970.2
XP_011517272.1
4
426
XM_017015067.1
XP_016870556.1
4
354
XM_017015069.1
XP_016870558.1
4
284
XM_017015070.1
XP_016870559.1
4
364
XM_017015071.1
XP_016870560.1
4
426
XM_017015072.1
XP_016870561.1
4
383
XM_017015073.1
XP_016870562.1
4
388
XM_017015074.1
XP_016870563.1
4
426
XM_017015077.1
XP_016870566.1
4
426
XM_017015078.1
XP_016870567.1
4
426
XM_017015079.1
XP_016870568.1
4
354
XM_017015080.1
XP_016870569.1
4
426
XM_017015081.1
XP_016870570.1
4
426
XM_017015082.1
XP_016870571.1
4
318
XM_017015083.1
XP_016870572.1
4
426
XM_017015084.1
XP_016870573.1
4
426
XM_017015085.1
XP_016870574.1
4
334
XM_017015086.1
XP_016870575.1
4
426
GenBank mRNA
AB033087.1
-
4
314
AK057236.1
-
4
401
AK296342.1
-
4
282
AK302709.1
-
4
288
AL162059.1
-
2
85
BC036369.1
-
4
453
BC045650.1
-
4
1253
5268902
BC059405.1
-
4
411
5296117
Back To Top
More Information
Panther Classification:
Molecular Function -
transcription cofactor
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
transcription, DNA-templated
biological_process
Wnt signaling pathway
beta-catenin-TCF complex assembly
molecular_function
chromatin binding
transcription factor activity, RNA polymerase II distal enhancer sequence-specific binding
transcription corepressor activity
protein binding
repressing transcription factor binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-8gqds:80/10.239.164.183:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0