Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC100910877 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003751127.3 | XP_003751175.1 | 11 | 1287 |
Gene Symbol: Cyp3a23/3a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013105.2 | NP_037237.2 | 1262 | |||
GenBank mRNA | BC088163.1 | - | 1262 | 7370410 | ||
D13912.1 | - | 11 | 1257 | |||
D29967.1 | - | 1259 | ||||
FQ211106.1 | - | 1272 | ||||
FQ218135.1 | - | 1993 | ||||
L24207.1 | - | 1249 | ||||
M10161.1 | - | 1270 | ||||
M28452.1 | - | 130 | ||||
X64401.1 | - | 1259 | ||||
X96721.1 | - | 1259 |