Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Camk2n1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173337.1 | NP_775459.1 | 1, 2 | 176 | ||
GenBank mRNA | AF271156.1 | - | 1, 2 | 176 |