Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gnptg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100493.1 | NP_001093963.1 | 8, 9 | 634 | ||
XM_006245935.2 | XP_006245997.1 | 8, 9 | 651 | |||
GenBank mRNA | BC098716.1 | - | 8, 9 | 616 | ||
BC158818.1 | - | 8, 9 | 634 | 7374209 | ||
FQ213320.1 | - | 8, 9 | 641 | |||
FQ213586.1 | - | 8, 9 | 599 | |||
FQ213702.1 | - | 8, 9 | 634 |