Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TM7SF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003273.3 | NP_003264.2 | 7, 8 | 1054 | ||
NR_102367.1 | 6, 7 | 999 | ||||
GenBank mRNA | AF023676.1 | - | 5, 6 | 1548 | ||
BC009052.1 | - | 7, 8 | 1020 | 3874367 |