Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cops3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004200.1 | NP_001004200.1 | 8 | 948 | ||
GenBank mRNA | BC079143.1 | - | 8 | 948 | 7128107 | |
FQ213575.1 | - | 4 | 482 | |||
FQ213666.1 | - | 4 | 479 | |||
FQ228882.1 | - | 8 | 989 |