Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Naa38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105794.2 | NP_001099264.2 | 2, 3 | 548 | ||
XM_008767795.1 | XP_008766017.1 | 2, 3 | 657 | |||
GenBank mRNA | FQ216987.1 | - | 2, 3 | 318 |