Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc16a11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105797.2 | NP_001099267.2 | 6 | 1199 | ||
XM_006246646.1 | XP_006246708.1 | 6 | 1339 | |||
XM_006246647.2 | XP_006246709.1 | 5 | 1882 | |||
XM_006246648.2 | XP_006246710.1 | 5 | 1184 | |||
XM_006246649.2 | XP_006246711.1 | 4 | 903 | |||
XM_008767805.1 | XP_008766027.1 | 5 | 1743 | |||
GenBank mRNA | BC169003.1 | - | 6 | 1339 | 7122746 | |
FQ219143.1 | - | 3 | 674 |