Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zmynd15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105800.1 | NP_001099270.1 | 5, 6 | 1672 | ||
XM_006246661.2 | XP_006246723.1 | 4, 5 | 1187 | |||
XM_006246662.2 | XP_006246724.1 | 4, 5 | 1174 |